Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX52 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536507
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536507 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536507 Supplier Thermo Scientific Supplier No. PA536507
Add to Cart
Add to Cart

DDX52 Polyclonal Antibody

DDX52 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX52
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.2
Gene DDX52
Gene Accession No. Q9Y2R4
Gene Alias 2700029C06Rik; ATP-dependent RNA helicase ROK1-like; ATP-dependent, RNA helicase; DDX52; DEAD (Asp-Glu-Ala-Asp) box polypeptide 52; DEAD box protein 52; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 52; DEAD-box helicase 52; HUSSY19; HUSSY-19; probable ATP-dependent RNA helicase DDX52; Rok1; rROK1L
Gene Symbols DDX52
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 553-598 of human DDX52.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11056
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.