Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX52 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | DDX52 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-4143
![]() |
Bethyl Laboratories, Inc
A303054AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp, are putative RNA helicases implicated in several cellular processes involving modifications of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family may be involved in embryogenesis, spermatogenesis, and cellular growth and division. DEAD box protein 52 (DDX52), also known as ATP-dependent RNA helicase ROK1-like or HUSSY-19, is a 599 amino acid protein belonging to the DEAD box helicase family. Localized to the nucleus, DDX52 is phosphorylated by ATM or ATR upon DNA damage. DDX52 contains one helicase ATP-binding domain and one helicase C-terminal domain.Specifications
DDX52 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
ATP dependent RNA helicase ROK1 like, ATP-dependent RNA helicase ROK1-like, DDX52, DDX52, DEAD (Asp-Glu-Ala-Asp) box polypeptide 52, DEAD box protein 52, Probable ATP dependent RNA helicase DDX52, Probable ATP-dependent RNA helicase DDX52, ROK1 | |
DDX52 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9Y2R4 | |
11056 | |
Between 549 and 599 | |
Primary | |
4° C | |
DDX52 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title