Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX52 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501564140
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4140 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4140 Supplier Bethyl Laboratories, Inc Supplier No. A303053AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Bovine

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp, are putative RNA helicases implicated in several cellular processes involving modifications of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family may be involved in embryogenesis, spermatogenesis, and cellular growth and division. DEAD box protein 52 (DDX52), also known as ATP-dependent RNA helicase ROK1-like or HUSSY-19, is a 599 amino acid protein belonging to the DEAD box helicase family. Localized to the nucleus, DDX52 is phosphorylated by ATM or ATR upon DNA damage. DDX52 contains one helicase ATP-binding domain and one helicase C-terminal domain.

Specifications

Antigen DDX52
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene DDX52
Gene Accession No. Q9Y2R4
Gene Alias ATP dependent RNA helicase ROK1 like, ATP-dependent RNA helicase ROK1-like, DDX52, DDX52, DEAD (Asp-Glu-Ala-Asp) box polypeptide 52, DEAD box protein 52, Probable ATP dependent RNA helicase DDX52, Probable ATP-dependent RNA helicase DDX52, ROK1
Gene Symbols DDX52
Host Species Rabbit
Immunogen Between 499 and 549
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11056
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.