Learn More
DDX52 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303053A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Bovine
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp, are putative RNA helicases implicated in several cellular processes involving modifications of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family may be involved in embryogenesis, spermatogenesis, and cellular growth and division. DEAD box protein 52 (DDX52), also known as ATP-dependent RNA helicase ROK1-like or HUSSY-19, is a 599 amino acid protein belonging to the DEAD box helicase family. Localized to the nucleus, DDX52 is phosphorylated by ATM or ATR upon DNA damage. DDX52 contains one helicase ATP-binding domain and one helicase C-terminal domain.Specifications
DDX52 | |
Polyclonal | |
Unconjugated | |
DDX52 | |
ATP dependent RNA helicase ROK1 like, ATP-dependent RNA helicase ROK1-like, DDX52, DDX52, DEAD (Asp-Glu-Ala-Asp) box polypeptide 52, DEAD box protein 52, Probable ATP dependent RNA helicase DDX52, Probable ATP-dependent RNA helicase DDX52, ROK1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11056 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9Y2R4 | |
DDX52 | |
Between 499 and 549 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.