Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DDX53 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA514403
Description
DDX53 Polyclonal Antibody for Western Blot, Flow

Specifications
DDX53 | |
Polyclonal | |
Unconjugated | |
DDX53 | |
CAGE; cancer/testis antigen 26; Cancer-associated gene protein; CT26; DDX53; DEAD (Asp-Glu-Ala-Asp) box polypeptide 53; DEAD box protein 53; DEAD box protein CAGE; DEAD-box helicase 53; DEAD-box protein CAGE; probable ATP-dependent RNA helicase DDX53 | |
Rabbit | |
Ammonium Sulfate Precipitation, Size-exclusion - Dialysis | |
RUO | |
168400 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q86TM3 | |
DDX53 | |
KLH conjugated synthetic peptide between 597-626 amino acids from the C-terminal region of human DDX53. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction