Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX53 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | DDX53 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
DDX53 Polyclonal specifically detects DDX53 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DDX53 | |
Polyclonal | |
Purified | |
RUO | |
Mouse | |
CAGEprobable ATP-dependent RNA helicase DDX53, Cancer/testis antigen 26, Cancer-associated gene protein, CT26EC 3.6.4.13, DEAD (Asp-Glu-Ala-Asp) box polypeptide 53, DEAD box protein 53, DEAD box protein CAGE, DEAD-box protein CAGE, EC 3.6.1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human DDX53 (NP_874358.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cancer | |
PBS with 50% glycerol, pH7.3. | |
168400 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title