Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX54 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060678B
Description
DDX54 Polyclonal antibody specifically detects DDX54 in Human, Mouse samples. It is validated for Western BlotSpecifications
DDX54 | |
Polyclonal | |
Western Blot | |
APR-5, ATP-dependent RNA helicase DP97, DEAD (Asp-Glu-Ala-Asp) box polypeptide 54, DEAD box helicase 97 KDa, DEAD box protein 54, DEAD box RNA helicase 97 kDa, DP97ATP-dependent RNA helicase DDX54, EC 3.6.1, EC 3.6.4.13, MGC2835 | |
The immunogen recognized by this antibody maps to a region between residue 800 and 850 of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 54 using the numbering given in entry NP_076977.3 (GeneID 79039). | |
0.1 mL | |
Signal Transduction | |
79039 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction