Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DDX54 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555805
Description
Immunogen sequence: VDTKLNEQLK TSFFLVREDT KAAVLLHLLH NVVRPQDQTV VFVATKHHAE YLTELLTTQR VSCAHIYSAL DPTARKINLA KFTLGKCSTL IVTDLAARGL DIPLLDNVIN YSFP Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The nucleolar protein encoded by this gene interacts in a hormone-dependent manner with nuclear receptors, and represses their transcriptional activity. Alternative splice variants that encode different isoforms have been found for this gene.
Specifications
DDX54 | |
Polyclonal | |
Unconjugated | |
DDX54 | |
2410015A15Rik; 5-Apr; AI414901; ATP-dependent RNA helicase; ATP-dependent RNA helicase DDX54; ATP-dependent RNA helicase DP97; chunp6913; Ddx54; DEAD (Asp-Glu-Ala-Asp) box polypeptide 54; DEAD box helicase 97 KDa; DEAD box protein 54; DEAD box RNA helicase 97 kDa; DEAD-box helicase 54; DP97; LOW QUALITY PROTEIN: ATP-dependent RNA helicase DDX54; mgc2835; RGD1562539; zgc:111908 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79039 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TDD1 | |
DDX54 | |
Recombinant protein corresponding to Human DDX54. Recombinant protein control fragment (Product #RP-89340). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction