Learn More
DDX54 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301314AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The nucleolar protein encoded by this gene interacts in a hormone-dependent manner with nuclear receptors, and represses their transcriptional activity. Alternative splice variants that encode different isoforms have been found for this gene. [provided by RefSeq, Jul 2008].Specifications
| DDX54 | |
| Polyclonal | |
| Unconjugated | |
| DDX54 | |
| APR-5, ATP-dependent RNA helicase DP97, DEAD (Asp-Glu-Ala-Asp) box polypeptide 54, DEAD box helicase 97 KDa, DEAD box protein 54, DEAD box RNA helicase 97 kDa, DP97, MGC2835 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79039 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q8TDD1 | |
| DDX54 | |
| Between 800 and 850 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.