Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX55 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116932
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116932 Supplier Invitrogen™ Supplier No. PA5116932
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, A-549, U-87MG, Mouse spleen Immunogen sequence: MEHVTEGSWE SLPVPLHPQV LGALRELGFP YMTPVQSATI PLFMRNKDVA AEAVTGSGKT LAFVIPILEI LLRREEKLKK SQVGAIIITP TRELAIQIDE VLSHFTKHFP EFSQILWIGG RNPGEDVERF KQQGGNIIVA TPGRLEDMFR RKAEGLDLAS CVRSLDVLVL DEADRLLDMG FEASINTILE FLPKQRRTGL FSATQTQEVE NLVRAGLRNP VRVSVKEKGV AASSAQKTPS RLENYYMVCK ADEKFNQLVH FLRNHKQEKH LVFFSTCACV EYYGKALEVL VKGVKIMCIH GKMKYKRNKI FMEFRKLQSG ILVCTDVMAR GIDIPEVNWV LQYDPPSNAS AFVHRCGRTA.

This gene encodes an enzyme which ubiquitinates proteins which participate in signaling pathways and apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX55
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DDX55
Gene Accession No. Q6ZPL9, Q8NHQ9
Gene Alias 2810021H22Rik; ATP-dependent RNA helicase DDX55; Ddx55; DEAD (Asp-Glu-Ala-Asp) box polypeptide 55; DEAD box protein 55; DEAD-box helicase 55; Kiaa1595; mKIAA1595
Gene Symbols DDX55
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-360 of human DDX55 (NP_0659871).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57696, 67848
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.