Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX55 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536431
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536431 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536431 Supplier Invitrogen™ Supplier No. PA536431
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 68 kDa. Purity is >95% by SDS-PAGE.

This gene encodes an enzyme which ubiquitinates proteins which participate in signaling pathways and apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX55
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene DDX55
Gene Accession No. Q8NHQ9
Gene Alias 2810021H22Rik; ATP-dependent RNA helicase DDX55; Ddx55; DEAD (Asp-Glu-Ala-Asp) box polypeptide 55; DEAD box protein 55; DEAD-box helicase 55; Kiaa1595; mKIAA1595
Gene Symbols DDX55
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 102-150 of human DDX55.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57696
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.