Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX56 Monoclonal Antibody (OTI2E5)
GREENER_CHOICE

Catalog No. PIMA526555
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526555 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526555 Supplier Invitrogen™ Supplier No. MA526555
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DDX56 Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The protein encoded by this gene shows ATPase activity in the presence of polynucleotides and associates with nucleoplasmic 65S preribosomal particles. This gene may be involved in ribosome synthesis, most likely during assembly of the large 60S ribosomal subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX56
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2E5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DDX56
Gene Accession No. Q9NY93
Gene Alias 2600001H07Rik; 61-kd nucleolar helicase; ATP-dependent 61 kDa nucleolar RNA helicase; D11Ertd619e; DDX21; DDX26; DDX56; DEAD (Asp-Glu-Ala-Asp) box helicase 56; DEAD (Asp-Glu-Ala-Asp) box polypeptide 56; DEAD box protein 21; DEAD box protein 56; DEAD-box helicase 56; DEAD-box RNA helicase; NOH61; nucleolar helicase of 61 kDa; nucleolar RNA helicase homolog; probable ATP-dependent RNA helicase DDX56; putative nucleolar RNA helicase; RH-II/Gu; RNA helicase II/Gu
Gene Symbols DDX56
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 323-547 of DDX56 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54606
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.