Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX59 Antibody (OTI2G1), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272229AF532
Description
DDX59 Monoclonal specifically detects DDX59 in Human samples. It is validated for Immunohistochemistry.Specifications
DDX59 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
DDX59 | |
Recombinant protein fragment corresponding to amino acids 1-303 of human DDX59 (NP_001026895) produced in E.coli. | |
0.1 mL | |
83479.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI2G1 | |
Immunohistochemistry | |
DEAD (Asp-Glu-Ala-Asp) box polypeptide 59, DEAD box protein 59, DKFZP564B1023, EC 3.6.1, EC 3.6.4.13, Zinc finger HIT domain-containing protein 5, ZNHIT5probable ATP-dependent RNA helicase DDX59 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction