Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX59 Monoclonal Antibody (OTI1F11)
GREENER_CHOICE

Catalog No. PIMA526437
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526437 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526437 Supplier Invitrogen™ Supplier No. MA526437
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DDX59 Monoclonal Antibody for Western Blot, IHC (P)

DDX59 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX59
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1F11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DDX59
Gene Accession No. Q5T1V6
Gene Alias 1210002B07Rik; 4833411G06Rik; DDX59; DEAD (Asp-Glu-Ala-Asp) box polypeptide 59; DEAD box protein 59; DEAD-box helicase 59; OFD5; Probable ATP-dependent RNA helicase DDX59; zinc finger HIT domain-containing protein 5; Znhit5
Gene Symbols DDX59
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-303 of DDX59 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83479
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.