Learn More
Invitrogen™ DDX59 Monoclonal Antibody (OTI2G1)

Description
DDX59 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | DDX59 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2G1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DDX59 |
| Gene Accession No. | Q5T1V6 |
| Gene Alias | 1210002B07Rik; 4833411G06Rik; DDX59; DEAD (Asp-Glu-Ala-Asp) box polypeptide 59; DEAD box protein 59; DEAD-box helicase 59; OFD5; Probable ATP-dependent RNA helicase DDX59; zinc finger HIT domain-containing protein 5; Znhit5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.