Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DDX59 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531563
Description
Recommended positive controls: 293T, HeLa, A375. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DDX59 is a protein coding gene.
Specifications
DDX59 | |
Polyclonal | |
Unconjugated | |
DDX59 | |
1210002B07Rik; 4833411G06Rik; DDX59; DEAD (Asp-Glu-Ala-Asp) box polypeptide 59; DEAD box protein 59; DEAD-box helicase 59; OFD5; Probable ATP-dependent RNA helicase DDX59; zinc finger HIT domain-containing protein 5; Znhit5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83479 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q5T1V6 | |
DDX59 | |
Recombinant protein encompassing a sequence within the N-terminus region of human DDX59. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction