Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX59 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531732
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531732 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531732 Supplier Invitrogen™ Supplier No. PA531732
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2, Neuro2A, GL261, C8D30. Predicted reactivity: Mouse (90%), Rat (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

DDX59 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX59
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene DDX59
Gene Accession No. Q5T1V6, Q9DBN9
Gene Alias 1210002B07Rik; 4833411G06Rik; DDX59; DEAD (Asp-Glu-Ala-Asp) box polypeptide 59; DEAD box protein 59; DEAD-box helicase 59; OFD5; Probable ATP-dependent RNA helicase DDX59; zinc finger HIT domain-containing protein 5; Znhit5
Gene Symbols DDX59
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DDX59. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67997, 83479
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.