Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX59 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561454
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561454 Supplier Thermo Scientific Supplier No. PA561454
Add to Cart
Add to Cart

DDX59 Polyclonal Antibody

DDX59 Polyclonal Antibody for Western Blot, ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX59
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DDX59
Gene Accession No. Q5T1V6
Gene Alias 1210002B07Rik; 4833411G06Rik; DDX59; DEAD (Asp-Glu-Ala-Asp) box polypeptide 59; DEAD box protein 59; DEAD-box helicase 59; OFD5; Probable ATP-dependent RNA helicase DDX59; zinc finger HIT domain-containing protein 5; Znhit5
Gene Symbols DDX59
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DDX59. Recombinant protein control fragment (Product #RP-99775).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83479
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.