Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX6 Antibody (6D8), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | DDX6 |
---|---|
Clone | 6D8 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
DDX6 Monoclonal antibody specifically detects DDX6 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
DDX6 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ATP-dependent RNA helicase p54, DEAD (Asp-Glu-Ala-Asp) box polypeptide 6, DEAD box protein 6, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 6 (RNA helicase, 54kD), EC 3.6.1, EC 3.6.4.13, FLJ36338, HLR2DEAD box-6, Oncogene RCK, probable ATP-dependent RNA helicase DDX6, RCKP54 | |
A synthesized peptide derived from Human DDX6 [UniProt P26196] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
6D8 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cancer, Tumor Suppressors | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
1656 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title