Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX6 Antibody (OTI2D6), Alexa Fluor™ 405, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270563AF405

Catalog No. N270563A405


Only null left
Add to Cart

Description

Description

DDX6 Monoclonal specifically detects DDX6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

DDX6
Monoclonal
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
DDX6
Recombinant protein fragment corresponding to amino acids 1-298 of human DDX6 (NP_004388) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG2a
Western Blot, Immunohistochemistry
OTI2D6
Western Blot, Immunohistochemistry
ATP-dependent RNA helicase p54, DEAD (Asp-Glu-Ala-Asp) box polypeptide 6, DEAD box protein 6, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 6 (RNA helicase, 54kD), EC 3.6.1, EC 3.6.4.13, FLJ36338, HLR2DEAD box-6, Oncogene RCK, probable ATP-dependent RNA helicase DDX6, RCKP54
Mouse
Affinity Purified
Cancer, Tumor Suppressors
1656.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.