Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DDX6 Recombinant Rabbit Monoclonal Antibody (6F9I6)
DDX6 Recombinant Rabbit Monoclonal Antibody (ARC1673)
Supplier: Invitrogen™ MA535825
Description
DDX6 Recombinant Monoclonal Antibody for Western Blot
This gene encodes a member of the DEAD box protein family. The protein is an RNA helicase found in P-bodies and stress granules, and functions in translation suppression and mRNA degradation. It is required for microRNA-induced gene silencing.
Specifications
DDX6 | |
Recombinant Monoclonal | |
0.53 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P26196, P54823 | |
DDX6 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 101-200 of human DDX6 (P26196). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot, Immunocytochemistry | |
6F9I6 | |
Unconjugated | |
DDX6 | |
1110001P04Rik; ATP-dependent RNA helicase p54; DDX6; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 6; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 6; DEAD (Asp-Glu-Ala-Asp) box helicase 6; DEAD (Asp-Glu-Ala-Asp) box polypeptide 6; DEAD box protein 6; DEAD box-6; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 6; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 6 (RNA helicase, 54kD); DEAD-box helicase 6; DEAD-box protein 6; E230023J21Rik; HLR2; mRCK/P54; Oncogene RCK; oncogene RCK homolog; p54; probable ATP-dependent RNA helicase DDX6; RCK; RGD1564560 | |
Rabbit | |
Affinity chromatography | |
RUO | |
13209, 1656, 500988 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction