Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX60 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281712IR
This item is not returnable.
View return policy
Description
DDX60 Polyclonal antibody specifically detects DDX60 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
DDX60 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
DyLight 755 | |
DEAD (Asp-Glu-Ala-Asp) box polypeptide 60, DEAD box protein 60 | |
DDX60 antibody was raised against a 17 amino acid synthetic peptide near the center of human DDX60. The immunogen is located within amino acids 1140 - 1190 of DDX60. Amino Acid Squence: LKKKQETKRPPKADKEA | |
0.1 mL | |
Innate Immunity | |
55601 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction