Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DDX60L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560278
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560278 Supplier Invitrogen™ Supplier No. PA560278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QEPHLNLGDS IRRDYEDLWN VVSHLVKEFN LGKSFPLRTT RRHFLRQEKS VIQEISLEKM PSVGFIPMTS AVIDEFVGDM MKDL Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 48%.

This gene encodes a member of the DExD/H-box helicase family of proteins, a subset of the super family 2 helicases. Members of the DExD/H-box helicase family share a conserved functional core comprised of two RecA-like globular domains. These domains contain conserved motifs that mediate ATP binding, ATP hydrolysis, nucleic acid binding, and RNA unwinding. In addition to functions in RNA metabolism, members of this family are involved in anti-viral immunity and act as cytosolic sensors of viral nucleic acids. The protein encoded by this gene has been shown to inhibit hepatitis C virus replication in response to interferon stimulation in cell culture. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX60L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DDX60L
Gene Accession No. Q5H9U9
Gene Alias DDX60L; DEAD (Asp-Glu-Ala-Asp) box polypeptide 60-like; DEAD box protein 60-like; DEAD-box helicase 60-like; probable ATP-dependent RNA helicase DDX60-like; putative ATP-dependent RNA helicase DDX60
Gene Symbols DDX60L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DDX60L. Recombinant protein control fragment (Product #RP-96766).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91351
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.