Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX60L Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2847640.1ML
Description
DDX60L Polyclonal specifically detects DDX60L in Human samples. It is validated for Western Blot.Specifications
| DDX60L | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 91351 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| probable ATP-dependent RNA helicase DDX60-like, DEAD (Asp-Glu-Ala-Asp) box polypeptide 60-like | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human DDX60L | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction