Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ De-tyrosinated alpha Tubulin Recombinant Rabbit Monoclonal Antibody (RM444)
GREENER_CHOICE

Catalog No. PIMA544591
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA544591 Supplier Invitrogen™ Supplier No. MA544591
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

De-tyrosinated alpha Tubulin Recombinant Monoclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen De-tyrosinated alpha Tubulin
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone RM444
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Gene TUBA1A
Gene Accession No. Q71U36
Gene Symbols TUBA1A
Host Species Rabbit
Immunogen A peptide corresponding to human dTyr a-tubulin. The immunogen range is a synthetic peptide within amino acids 351-451 of human alpha Tubulin which lacks a carboxy-terminal tyrosine group.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7846
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.