Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DEAF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583138
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583138 Supplier Invitrogen™ Supplier No. PA583138
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LIVVHTDGSI VETTGLKGPA APLTPGPQSP PTPLAPGQEK GGTKYNWDPS VYDSELPVRC RNISGTLYKN RLGSGGRGRC IKQGENWYSP TE Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.

HNF1A is a transcription factor required for the expression of several liver-specific genes. This protein functions as a homodimer and binds to the inverted palindrome 5'-GTTAATNATTAAC-3'.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEAF1
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Deaf1
Gene Accession No. O75398
Gene Alias AU042387; C230009B13Rik; DEAF1; DEAF-1 related transcriptional regulator (NUDR); DEAF1 transcription factor; DEAF1, transcription factor; deformed epidermal autoregulatory factor 1 (Drosophila); deformed epidermal autoregulatory factor 1 homolog; MRD24; Nuclear DEAF-1-related transcriptional regulator; NUDR; SPN; suppressin; Zinc finger MYND domain-containing protein 5; ZMYND5
Gene Symbols Deaf1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DEAF1. Recombinant protein control fragment (Product #RP-95534).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10522
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.