Learn More
Stoelting™ DeBakey Bulldog Clamp
Stoelting™ DeBakey Bulldog Clamp is useful in surgeries that require precise blood stoppage.
$730.50 - $772.00
| Catalog Number | Mfr. No. | Length (English) | Model | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Length (English) | Model | Price | Quantity & Availability | ||||
10-001-391
![]() |
Stoelting™
5212059 |
4.75 in. | Straight |
Each for $730.50
|
|
||||
10-001-392
![]() |
Stoelting™
5212060 |
4.75 in. | Curved |
Each for $730.50
|
|
||||
10-001-385
![]() |
Stoelting™
5212051 |
3 in. | Straight |
Each for $763.50
|
|
||||
10-001-386
![]() |
Stoelting™
5212052 |
3 in. | Curved |
Each for $772.00
|
|
||||
Description
One of the most common applications where a bulldog clamp can be used is during coronary surgery where blood flow must be stopped. In general the bulldog clamp, with its many angled varieties and surgical applications, is an essential tool during surgery where blood flow to an organ must be stopped or controlled.Specifications
| Neuroscience |