Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stoelting™ DeBakey Tissue Forceps
Shop All Stoelting Co Products

Click to view available options
Length (English):
6 in.
7.5 in.
8 in.
Width (Metric):
1.5 mm tip
2.5 mm tip
Description
Design minimizes the potential of damage (atraumatic)
Specifications
Specifications
For Use With (Application) | Neuroscience |
Length (English) | 6 in. |
Width (Metric) | 1.5 mm tip |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction