Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Deblock trichloroacetic acid soln., 3% w/w in dichloromethane
Click to view available options
Quantity:
200 mL
450 mL
2.5 L
Description
Solvent specially purified and analyzed for biotechnology applications, including peptide and oligonucleotide synthesis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solvent specially purified and analyzed for biotechnology applications, including peptide and oligonucleotide synthesis.

Specifications
Specifications
Chemical Name or Material | Deblock trichloroacetic acid soln. |
Color | Colorless |
MDL Number | MFCD00004177 |
UN Number | UN2810 |
Concentration | 3% w/w in dichloromethane |
Density | 1.325 |
Synonym | DEBLK soln. |
Quantity | 450 mL |
Physical Form | Liquid |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction