Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEC-205/CD205 Antibody (019), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP312780MFV500
Description
DEC-205/CD205 Monoclonal antibody specifically detects DEC-205/CD205 in Mouse samples. It is validated for Western BlotSpecifications
| DEC-205/CD205 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Mouse | |
| Purified |
| Western Blot | |
| 019 | |
| Western Blot | |
| CD205;DCL-1 fusion;DEC-205;gp200;Ly-75;Lymphocyte antigen 75;membrane antigen MN;RP23-10L17.4 | |
| This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the N-terminus of the Mouse DEC-205/CD205 | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction