Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEC-205/CD205 Antibody (MG38), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200328G0.1ML
Description
DEC-205/CD205 Monoclonal specifically detects DEC-205/CD205 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
DEC-205/CD205 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
The immunogen for this antibody is CD205 (DEC-205). | |
0.1 mL | |
Primary | |
Store at 4C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
MG38 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction