Learn More
DEC-205/CD205 Antibody (MG38), PE, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP200328PE0.1ML
Catalog No. NBP2003287
Only null left
Add to Cart
Description
Description
DEC-205/CD205 Monoclonal specifically detects DEC-205/CD205 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
DEC-205/CD205
Monoclonal
PE
PBS with 0.05% Sodium Azide
The immunogen for this antibody is CD205 (DEC-205).
0.1 mL
Primary
Store at 4C in the dark.
Flow Cytometry
MG38
Flow Cytometry
Mouse
Affinity Purified
RUO
Human
IgG2b κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only