Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEC-205/CD205 Antibody (NLDC-145), Alexa Fluor™ 546, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals DDX0020A546100
Description
DEC-205/CD205 Monoclonal specifically detects DEC-205/CD205 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
DEC-205/CD205 | |
Monoclonal | |
0.5mg/mL | |
Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:2000 | |
LY75 | |
Dec205/CD205 (NLDC145) was used as immunogen. | |
0.1 mg | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
NLDC-145 | |
Alexa Fluor 546 | |
NP_005840 | |
Rat | |
Ion exchange chromatography | |
RUO | |
Mouse | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction