Learn More
DEC1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300649A
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
Differentiated embryo-chondrocyte expressed gene 1 (DEC1) is an important protein involved in embryonic cell differentiation, proliferation, and apoptosis. It has also recently been shown to be regulated by hypoxia. The loss of DEC1 expression may be an early event in the development of lung cancer, while DEC1 may be up-regulated by hypoxia in other cancers and in more extreme hypoxia it may have a role in cell death.Specifications
| DEC1 | |
| Polyclonal | |
| Unconjugated | |
| BHLHE40 | |
| DEC-1, Differentially Expressed in Chondrocytes, Differentiated Embryo Chrondrocyte Expressed Gene 1, BHLHB2, Basic Helix-Loop-Helix Domain Containing | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8553 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| O14503 | |
| BHLHE40 | |
| Between 350 and C-term | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.