Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEC2/SHARP1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP119613AF488
Description
Specifically detects DEC2/SHARP1 in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting
DEC2/SHARP1 Polyclonal specifically detects DEC2/SHARP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DEC2/SHARP1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Q9C0J9 | |
BHLHE41 | |
A synthetic peptide made to an internal portion of the human DEC2/SHARP1 inbetween residues 1-75 [UniProt Q9C0J9]. | |
Affinity Purified | |
RUO | |
79365 | |
Store at 4C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
basic helix-loop-helix domain containing, class B, 3, basic helix-loop-helix family, member e41, bHLHb3, BHLHB3class E basic helix-loop-helix protein 41, bHLHe41, Class B basic helix-loop-helix protein 3, DEC2Enhancer-of-split and hairy-related protein 1, Differentially expressed in chondrocytes protein 2, SHARP-1, SHARP1hDEC2 | |
Rabbit | |
50 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction