Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEC2/SHARP1 Antibody (OTI5B1), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272203IR
Description
DEC2/SHARP1 Monoclonal specifically detects DEC2/SHARP1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
DEC2/SHARP1 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BHLHE41 | |
Recombinant protein fragment corresponding to amino acids 1-297 of human DEC2/SHARP1 (NP_110389) produced in E.coli. | |
0.1 mL | |
79365.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI5B1 | |
Western Blot, Immunohistochemistry | |
basic helix-loop-helix domain containing, class B, 3, basic helix-loop-helix family, member e41, bHLHb3, BHLHB3class E basic helix-loop-helix protein 41, bHLHe41, Class B basic helix-loop-helix protein 3, DEC2Enhancer-of-split and hairy-related protein 1, Differentially expressed in chondrocytes protein 2, SHARP-1, SHARP1hDEC2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction