Learn More
Decahydronaphthalene, cis + trans, 98%
Description
Decahydronaphthalene is widely used as an industrial solvent for resins and fuel additives. It is a substitute for turpentine in lacquers, shoe polishes and waxes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Decahydronaphthalene is widely used as an industrial solvent for resins and fuel additives. It is a substitute for turpentine in lacquers, shoe polishes and waxes.
Solubility
Miscible with acetone, benzene, ether, methanol, aniline, decalin, chloroform, butanol and ethanol. Immiscible with water.
Notes
Store in cool place. Air, light and heat sensitive. Heat and light accelerate peroxide formation. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | <-45°C |
| Density | 0.88 |
| Boiling Point | 189°C to 191°C |
| Flash Point | 57°C (134°F) |
| Odor | Mild |
| Refractive Index | 1.474 |
| Quantity | 10,000 mL |
| UN Number | UN1147 |
| Beilstein | 878165 |
| Merck Index | 14,2846 |
| Show More |
Safety and Handling
| Hazard Category | H226-H304-H314-H331-H335 |
| Hazard Statement | GHS H Statement H331-H226-H315-H319-H335 Toxic if inhaled. Flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DECAHYDRONAPHTHALENE |
| EINECSNumber | 202-046-9 |
| RTECSNumber | QJ3150000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.