Learn More
StatLab™ Decal Decalcifier
GSA_VASDP
Catalog No. p-7227722
Click to view available options
Quantity:
4 x 1 gal.
4 x 32 oz. Bottle
5 gal. Cube
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STL121132
Supplier: StatLab™ 121132
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
A gentle decalcifier formulated from semiconductor grade hydrochloric acid, EDTA, and highly purified reagent grade water.
  • Can be used to safely decalcify all types of tissue samples, from needle biopsies to cortical bone
  • Mild enough that most samples can be left in overnight
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 4 x 32 oz. Bottle
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.