Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        Decalcifying Solution, Krajian, for Decalcification of Bones, Ricca Chemical
 
    
    
    
        
            Shop All Ricca Chemical Company Products
        
    
    
    
    
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                500 mL
                            
                        
                            
                                1 L
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Natural Poly Bottle
                            
                        
                    Specifications
Specifications
| Physical Form | Liquid | 
| Chemical Name or Material | Decalcifying Solution | 
| Name Note | Krajian | 
| CAS | 7732-18-5, 64-18-6, 6132-04-3 | 
| CAS Min % | 70.0, 21.27, 6.98 | 
| CAS Max % | 73.0, 22.14, 7.26 | 
| Packaging | Natural Poly Bottle | 
| Color | Colorless | 
| Quantity | 500 mL | 
| Grade | Laboratory | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                