Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decamethonium Bromide 98.0+%, TCI America™
SDP

Catalog No. D131425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D131425G 25 g
D13145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D131425G Supplier TCI America Supplier No. D131425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 541-22-0
Molecular Formula C16H38Br2N2
Molecular Weight (g/mol) 418.302
MDL Number MFCD00011779
InChI Key HLXQFVXURMXRPU-UHFFFAOYSA-L
Synonym decamethonium bromide, decamethonium dibromide, decacuran, syncurine, decamethylene bis trimethylammonium bromide, dekamethoniumbromid czech, decametonio bromuro dcit, unii-55c6rk944k, decamethylenebis trimethylammonium bromide
PubChem CID 10921
IUPAC Name trimethyl-[10-(trimethylazaniumyl)decyl]azanium;dibromide
SMILES C[N+](C)(C)CCCCCCCCCC[N+](C)(C)C.[Br-].[Br-]

Specifications

Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 418.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Decamethonium Bromide
EINECSNumber (2)-0175
RTECSNumber BP5950000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.