Learn More
Decamethylferrocene, 99%, Thermo Scientific Chemicals
Description
Decamethylferrocene is used as a superior internal reference redox standard to ferrocene in voltammetric and amperometric titrations. It is used as photocathode for the detection of barium fluoride scintillation light.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 298°C to 300°C (sublimation) |
Odor | Odorless |
Quantity | 5 g |
Sensitivity | Air and moisture sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 326.31 |
Percent Purity | 99% |
Physical Form | Crystalline |
Chemical Name or Material | Decamethylferrocene |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.