Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decane, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187756
Click to view available options
Quantity:
5 mL
25 mL

Decane is an alkane hydrocarbon. It exists as a colourless flammable liquid. It is mostly used as solvent. Its vapors may be narcotic at high concentrations.

Petroleum
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 410°F
CAS 124-18-5
Vapor Pressure 3.77 mmHg (37.7°C)
Vapor Density 4.9 (vs. air)
Density 0.73 g/mL (at 25°C)
Flash Point 57°C
Physical Form Neat
Linear Formula CH3(CH2)8 CH3
MDL Number MFCD00008954
Melting Point -30°C
Boiling Point 174°C
Type Analytical Standard
Quantity 5 mL
Formula Weight 142.28
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.