Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decanoic Anhydride 98.0+%, TCI America™
SDP

Catalog No. C0020100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0020100G 100 g
C002025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0020100G Supplier TCI America Supplier No. C0020100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2082-76-0
Molecular Formula C20H38O3
Molecular Weight (g/mol) 326.521
MDL Number MFCD00010460
InChI Key HTWWKYKIBSHDPC-UHFFFAOYSA-N
Synonym decanoic anhydride, capric anhydride, n-capric anhydride, n-decanoic anhydride, decanoic acid anhydride, acmc-209few, decanoic acid, anhydride, decanoic acid,1,1'-anhydride, htwwkykibshdpc-uhfffaoysa
PubChem CID 74960
IUPAC Name decanoyl decanoate
SMILES CCCCCCCCCC(=O)OC(=O)CCCCCCCCC

Specifications

Color White
Quantity 100 g
UN Number 3261
Formula Weight 326.52
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Decanoic Anhydride
EINECSNumber (2)-3916
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.