Learn More
Corning™ DeckWorks™ Tip Stations, Sterile

Description
Corning DeckWorks tip station maximizes bench space while minimizing plastic waste. Tip stations contain 960 tips, are available in extended-length 10 μL and traditional 200 μL sizes, and are the ideal single and multi-channel pipetting stations. The 5-layer deck design (2 x 96) matches the microtiter plate format. This stable pipetting platform fits easily into sterile hood environments. Lids can be snap-positioned to expose a single side (96 tips) only. Applications include pre- and post-PCR procedures, cell culture, and assays requiring large tip quantities. Corning DeckWorks tip stations are offered both sterile and nonsterile.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Volume (Metric) | 1 to 200 μL |
Sterility | Sterile |
Color | Natural |
Tip Style | Round |
Packaging | Multi-pack |
For Use With (Equipment) | Universal Fit, Multichannel Pipetting Stations |
Graduated | Yes |
Product Line | DeckWorks |
Material | Polypropylene |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.