Learn More
VAI DECON-CYCLE™ II Germicidal Detergent
DECON-CYCLE™ II Germicidal Detergent is a low pH phenolic that is effective as hard non-porous surface disinfectant/detergent formulated for effective aseptic and controlled environment sanitation.
$416.50 - $929.50
Specifications
Sterility | Sterile |
---|---|
For Use With (Application) | Kills broad spectrum of pathogenic Bacteria, pharmaceutical, biotechnology, medical device and health care industries |
Catalog Number | Mfr. No. | Volume (Metric) | Dilution | Format | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Dilution | Format | Price | Quantity & Availability | |||
19-546-601
|
VAI
DCY20616Z02 |
473 mL | SimpleMix |
Case of 12 for $744.50
|
![]() |
||||
19-546-528
|
VAI
DCY20616Z01 |
473 mL | 1:256 | SimpleMix |
Case of 12 for $929.50
|
|
|||
19-546-598
|
VAI
DCY2041Z |
1:128 |
Case of 4 for $416.50
|
|
|||||
19-546-597
|
VAI
DCY202 |
3.8 L | 1:256 | Concentrate |
Case of 4 for $920.50
|
![]() |
|||
Description
A low pH, dilutable, hospital detergent effective in 10 minutes at 20°C in hard water up to 400 ppm (calculated as CaCO3) in the presence of 5% blood serum. Recommended for use in pharmaceutical, biotechnology, medical device manufacturing, and in healthcare institutions to control cross-contamination hazards.
- Multi-phenolic formula designed to clean, disinfect, and deodorize any washable inanimate surface.
- Filtered at 0.2 μm and gamma irradiated.
- Strong enough to kill a broad spectrum of pathogenic bacteria, yet mild enough to have no harmful effect on disinfected surfaces.
- Completely traceable and validated for sterility and shelf life.
- Filled in an ISO 5 (Grade A/B, Former Class 100).
- Lot sterility tested according to current USP compendium.
- Completely traceable from start to finish.
Order Info
Double-bag packaging.
Compliance
Completely tested according to current USP compendium.
Specifications
Sterile | |
Kills broad spectrum of pathogenic Bacteria, pharmaceutical, biotechnology, medical device and health care industries |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.