Learn More
VAI DECON-CYCLE™ II Germicidal Detergent, Sterile
Shop All VAI ProductsDescription
Includes
Lot-specific Certificate of Irradiation, Certificate of Analysis, LAL testing analysis and sterility report
A low pH, dilutable, hospital detergent effective in 10 minutes at 20°C in hard water up to 400 ppm (calculated as CaCO3) in the presence of 5% blood serum. Recommended for use in pharmaceutical, biotechnology, medical device manufacturing, and in healthcare institutions to control cross-contamination hazards.
- Multi-phenolic formula designed to clean, disinfect, and deodorize any washable inanimate surface
- Filtered at 0.2 μm and gamma irradiated
- Strong enough to kill a broad spectrum of pathogenic bacteria, yet mild enough to have no harmful effect on disinfected surfaces
- Completely traceable and validated for sterility and shelf life
- Filled in an ISO 5 (Grade A/B, Former Class 100)
- Lot sterility tested according to current USP compendium
- Completely traceable from start to finish
Order Info
Double-bag packaging.
Compliance
Completely tested according to current USP compendium.
Specifications
Specifications
Sterility | Sterile |
Dilution | 1:256 |
Format | Concentrate |
For Use With (Application) | Kills Broad Spectrum of Pathogenic Bacteria, Pharmaceutical, Biotechnology, Medical Device and Health Care Industries |
Volume (English) | 2 oz. |
Volume (Metric) | 59 mL |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.