Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Decon™ LeLoop™ Inoculating Loops

Click to view available options
Volume:
1 μL
10 μL
2.45 μL
3.41 μL
5.45 μL
Calibrated:
No
Yes
Diameter (Metric) Inside:
0.14 cm
0.22 cm
0.29 cm
0.35 cm
0.4 cm
Description
More economical than platinum or disposable plastic loops
Reusable loops are made of high-quality twisted nichrome wire.
- Loops slide smoothly across agar surfaces without tearing or gouging
- The nichrome wire cools rapidly, saving time
- Loops are available calibrated or noncalibrated
- Calibrated in accordance with ASM/APHA via Evans Blue Test and include calibration certificates
- LeLoop™ loops come without holders, which are available separately
Specifications
Specifications
Sterility | Non-sterile |
Length (English) | 3 in. |
Length (Metric) | 7.62 cm |
Volume | 1 μL |
Diameter (Metric) Inside | 0.14 cm |
Material | Nichrome |
Diameter (English) Inside | 0.05 in. |
Size | Small |
Product Type | Inoculating Loop |
Includes | Calibration certificates in accordance with ASM/APHA via evans blue test |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction