Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Decorin Antibody (DCN/6289), Novus Biologicals™

Mouse Monoclonal Antibody
$216.50 - $443.50
Specifications
Antigen | Decorin |
---|---|
Clone | DCN/6289 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL |
Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
Decorin Monoclonal antibody specifically detects Decorin in Human, Mouse, Porcine, Bovine, Canine, Chicken, Equine, Elk, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Decorin | |
0.2 mg/ml | |
Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer | |
10mM PBS with 0.05% BSA | |
1634 | |
IgG1 λ | |
Protein A or G purified |
DCN/6289 | |
Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Pig, Bovine, Canine | |
Bone proteoglycan II, CSCD, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-S2, proteoglycan core protein, SLRR1BPGS2, small leucine-rich protein 1B | |
Recombinant full-length Decorin protein | |
Primary | |
Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title