Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DECR1 Monoclonal Antibody (OTI16C2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA808966

Description
This gene encodes an accessory enzyme which participates in the beta-oxidation and metabolism of unsaturated fatty enoyl-CoA esters.Specifications
DECR1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q16698 | |
DECR1 | |
Full length human recombit protein of human DECR1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI16C2 | |
Unconjugated | |
DECR1 | |
DECR, NADPH, SDR18C1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1666 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction