Learn More
DECR1 Mouse anti-Human, Clone: OTI5D6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808947

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes an accessory enzyme which participates in the beta-oxidation and metabolism of unsaturated fatty enoyl-CoA esters.Specifications
DECR1 | |
Monoclonal | |
Unconjugated | |
DECR1 | |
DECR, NADPH, SDR18C1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1666 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI5D6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q16698 | |
DECR1 | |
Full length human recombit protein of human DECR1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.